Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTTTCTCTCCTGTGTGAATTCT[C/G]TGGTGTTGAATAAGGCATGTTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF287 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF287 - zinc finger protein 287 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020653.2 | 2622 | Missense Mutation | CAC,CAG | H,Q 723 | NP_065704.2 | |
XM_011523968.2 | 2622 | Missense Mutation | CAC,CAG | H,Q 723 | XP_011522270.1 | |
XM_017024887.1 | 2622 | Missense Mutation | CAC,CAG | H,Q 723 | XP_016880376.1 |