Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAACTGGCCCGGGGCCTTTTGCT[G/T]CTGGCTTCCGACTCCAGGGTTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615498 MIM: 601243 | ||||||||||||||||||||
Literature Links: |
MIEF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIEF2 - mitochondrial elongation factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOP3A - topoisomerase (DNA) III alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320759.1 | 3203 | Missense Mutation | AGA,AGC | R,S 868 | NP_001307688.1 | |
NM_004618.4 | 3203 | Missense Mutation | AGA,AGC | R,S 963 | NP_004609.1 | |
XM_011524001.2 | 3203 | Missense Mutation | AGA,AGC | R,S 493 | XP_011522303.1 |