Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGCTGTTAAAGCCAGGAGGCCA[A/G]ATTCCTCAGATGGGACTCCAGTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614515 MIM: 611850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR179 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR179 - G protein-coupled receptor 179 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004334.3 | 6782 | Missense Mutation | TCT,TTT | S,F 2254 | NP_001004334.3 |
MRPL45 - mitochondrial ribosomal protein L45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |