Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142961 MIM: 142962 MIM: 142963 MIM: 142964 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXB-AS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXB-AS3 - HOXB cluster antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB6 - homeobox B6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB7 - homeobox B7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB8 - homeobox B8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024016.3 | 6025 | Silent Mutation | ACA,ACC | T,T 158 | NP_076921.1 | |
XM_005257286.2 | 6025 | Silent Mutation | ACA,ACC | T,T 157 | XP_005257343.1 | |
XM_017024564.1 | 6025 | Silent Mutation | ACA,ACC | T,T 158 | XP_016880053.1 |
HOXB9 - homeobox B9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |