Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCTCGCCGTCGCCCACCGGTC[C/T]CTCGGCGGCCGCCATCTTGGCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP88 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP88 - nucleoporin 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320653.1 | 28 | Missense Mutation | NP_001307582.1 | |||
NM_002532.5 | 28 | Missense Mutation | NP_002523.2 | |||
XM_011523893.2 | 28 | UTR 5 | XP_011522195.1 | |||
XM_017024694.1 | 28 | UTR 5 | XP_016880183.1 |
RPAIN - RPA interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033002.3 | 28 | Intron | NP_001028174.2 | |||
NM_001160243.1 | 28 | Intron | NP_001153715.1 | |||
NM_001160244.1 | 28 | Intron | NP_001153716.1 | |||
NM_001160246.1 | 28 | Intron | NP_001153718.1 | |||
NM_001160266.1 | 28 | Intron | NP_001153738.1 |