Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGCCCTGGACCCTGGCTGACCG[C/G]TCAGGTTGGCGTAATCAATCCTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606538 | ||||||||||||||||||||
Literature Links: |
MYO1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO1C - myosin IC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080779.1 | 2898 | Missense Mutation | ACC,AGC | T,S 965 | NP_001074248.1 | |
NM_001080950.1 | 2898 | Missense Mutation | ACC,AGC | T,S 946 | NP_001074419.1 | |
NM_033375.4 | 2898 | Missense Mutation | ACC,AGC | T,S 930 | NP_203693.3 |