Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCATTCTTAAGAACCTGGACCC[A/G]GCTCTCCTCACCAGCGTCTCCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610067 MIM: 609517 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO18A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO18A - myosin XVIIIA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_078471.3 | 1483 | UTR 3 | NP_510880.2 | |||
NM_203318.1 | 1483 | UTR 3 | NP_976063.1 |
TIAF1 - TGFB1-induced anti-apoptotic factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004740.3 | 1483 | Missense Mutation | CGG,TGG | R,W 25 | NP_004731.2 |