Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCATCTGAAGACTGAGACAGC[A/G]TGTGGTAGGGGTAAGAACATTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDK2 - pyruvate dehydrogenase kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAMD14 - sterile alpha motif domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257359.1 | 2237 | Missense Mutation | ACG,ATG | T,M 307 | NP_001244288.1 | |
NM_174920.3 | 2237 | Missense Mutation | ACG,ATG | T,M 335 | NP_777580.1 | |
XM_011524490.2 | 2237 | Missense Mutation | ACG,ATG | T,M 90 | XP_011522792.1 | |
XM_017024322.1 | 2237 | Missense Mutation | ACG,ATG | T,M 390 | XP_016879811.1 | |
XM_017024323.1 | 2237 | Missense Mutation | ACG,ATG | T,M 307 | XP_016879812.1 | |
XM_017024324.1 | 2237 | Missense Mutation | ACG,ATG | T,M 335 | XP_016879813.1 |