Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAAACCAGACGGAGAAGCTGCG[G/T]CCATTCCACTGCAGGCAGCAGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611041 | ||||||||||||||||||||
Literature Links: |
TRIM47 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM47 - tripartite motif containing 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033452.2 | 1605 | Silent Mutation | GGA,GGC | G,G 526 | NP_258411.2 | |
XM_005257787.4 | 1605 | Silent Mutation | GGA,GGC | G,G 288 | XP_005257844.1 | |
XM_005257788.4 | 1605 | Silent Mutation | GGA,GGC | G,G 288 | XP_005257845.1 |
TRIM65 - tripartite motif containing 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |