Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCAGTATCACTGCCATCTGAC[C/T]CCTTCAAACATGTCTGATGAAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSF5 - heat shock transcription factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTMR4 - myotubularin related protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004687.4 | 3451 | Missense Mutation | GAG,GGG | E,G 1075 | NP_004678.3 | |
XM_005257784.2 | 3451 | Missense Mutation | GAG,GGG | E,G 1089 | XP_005257841.1 | |
XM_005257785.4 | 3451 | Missense Mutation | GAG,GGG | E,G 1079 | XP_005257842.1 | |
XM_005257786.4 | 3451 | Missense Mutation | GAG,GGG | E,G 1075 | XP_005257843.1 | |
XM_006722168.3 | 3451 | Missense Mutation | GAG,GGG | E,G 1075 | XP_006722231.1 | |
XM_011525460.2 | 3451 | Missense Mutation | GAG,GGG | E,G 1079 | XP_011523762.1 |