Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCGATCTCCAGCTCCTGCATC[A/G]TGCGTCTCAGGTCTGTGATCTCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 148030 MIM: 148020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT15 - keratin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002275.3 | 1241 | Missense Mutation | ACG,ATG | T,M 328 | NP_002266.2 | |
XM_011524784.2 | 1241 | Missense Mutation | ACG,ATG | T,M 335 | XP_011523086.1 | |
XM_017024614.1 | 1241 | Missense Mutation | ACG,ATG | T,M 335 | XP_016880103.1 |
KRT19 - keratin 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6510 - microRNA 6510 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |