Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTCGCAAGAGATGTAGGAAGGC[C/T]GCTTCAGAGGTTGTAGCTCACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf75 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf75 - chromosome 17 open reading frame 75 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022344.3 | Intron | NP_071739.2 | ||||
XM_005258022.3 | Intron | XP_005258079.1 | ||||
XM_017024940.1 | Intron | XP_016880429.1 | ||||
XM_017024941.1 | Intron | XP_016880430.1 |
RHBDL3 - rhomboid like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |