Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATCCCCTTAGGTAGCAAAGGAC[A/G]TGACACTTCATCAGGCCTTGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607954 MIM: 610818 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RANGRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RANGRF - RAN guanine nucleotide release factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177801.1 | 502 | Missense Mutation | ATG,GTG | M,V 122 | NP_001171272.1 | |
NM_001177802.1 | 502 | Missense Mutation | CAT,CGT | H,R 151 | NP_001171273.1 | |
NM_016492.4 | 502 | Missense Mutation | ATG,GTG | M,V 122 | NP_057576.2 | |
XM_005256618.4 | 502 | Intron | XP_005256675.1 |
SLC25A35 - solute carrier family 25 member 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320870.1 | 502 | Intron | NP_001307799.1 | |||
NM_001320871.1 | 502 | Intron | NP_001307800.1 | |||
NM_001320872.1 | 502 | Intron | NP_001307801.1 | |||
NM_001320877.1 | 502 | Intron | NP_001307806.1 | |||
NM_201520.2 | 502 | UTR 3 | NP_958928.1 | |||
XM_005256641.2 | 502 | Intron | XP_005256698.1 | |||
XM_011523846.1 | 502 | Intron | XP_011522148.1 | |||
XM_017024633.1 | 502 | Intron | XP_016880122.1 | |||
XM_017024634.1 | 502 | Intron | XP_016880123.1 |