Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACTCCCACTTGGGTTCCTTTGT[A/G]GTATGAAATAGAGACACTTCTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0753 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0753 - KIAA0753 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014804.2 | 3876 | UTR 3 | NP_055619.2 | |||
XM_006721612.2 | 3876 | UTR 3 | XP_006721675.1 | |||
XM_011524090.2 | 3876 | UTR 3 | XP_011522392.1 | |||
XM_011524091.2 | 3876 | UTR 3 | XP_011522393.1 | |||
XM_011524095.2 | 3876 | UTR 3 | XP_011522397.1 | |||
XM_011524096.1 | 3876 | Intron | XP_011522398.1 | |||
XM_011524098.2 | 3876 | UTR 3 | XP_011522400.1 | |||
XM_011524099.2 | 3876 | UTR 3 | XP_011522401.1 | |||
XM_011524100.2 | 3876 | UTR 3 | XP_011522402.1 | |||
XM_017025455.1 | 3876 | UTR 3 | XP_016880944.1 | |||
XM_017025456.1 | 3876 | UTR 3 | XP_016880945.1 | |||
XM_017025457.1 | 3876 | UTR 3 | XP_016880946.1 |