Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGACGCGGGGGTGGAAGTCTT[C/T]GTGCCCGTGCAAAGCGGCCTCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610917 MIM: 602326 | ||||||||||||||||||||
Literature Links: |
NARR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NARR - nine-amino acid residue-repeats | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256281.1 | 712 | Missense Mutation | CAA,CGA | Q,R 177 | NP_001243210.1 |
PROCA1 - protein interacting with cyclin A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB34 - RAB34, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142624.2 | 712 | Missense Mutation | AAA,GAA | K,E 87 | NP_001136096.2 | |
NM_001142625.2 | 712 | Missense Mutation | AAA,GAA | K,E 87 | NP_001136097.2 | |
NM_001144942.1 | 712 | Missense Mutation | AAA,GAA | K,E 30 | NP_001138414.1 | |
NM_001144943.1 | 712 | Missense Mutation | AAA,GAA | K,E 87 | NP_001138415.1 | |
NM_001256276.1 | 712 | Missense Mutation | AAA,GAA | K,E 30 | NP_001243205.1 | |
NM_001256277.1 | 712 | Missense Mutation | AAA,GAA | K,E 30 | NP_001243206.1 | |
NM_001256278.1 | 712 | Intron | NP_001243207.1 | |||
NM_031934.5 | 712 | Missense Mutation | AAA,GAA | K,E 30 | NP_114140.4 |
RPL23A - ribosomal protein L23a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD42A - small nucleolar RNA, C/D box 42A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD42B - small nucleolar RNA, C/D box 42B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD4A - small nucleolar RNA, C/D box 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD4B - small nucleolar RNA, C/D box 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLCD1 - TLC domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |