Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCAGAACCTCTTCCTCTCCCC[A/G]TGGAGCATCTCGTCCACCATGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173390 | ||||||||||||||||||||
Literature Links: |
SERPINB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINB2 - serpin family B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143818.1 | 171 | Silent Mutation | CCA,CCG | P,P 32 | NP_001137290.1 | |
NM_002575.2 | 171 | Silent Mutation | CCA,CCG | P,P 32 | NP_002566.1 |