Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGACTGGGCTTTCTGAAAAGA[C/G]TTCTGTCTGTAGGCAAGTAGACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM210A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM210A - family with sequence similarity 210 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNMT - RNA guanine-7 methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308263.1 | 381 | Missense Mutation | ACT,AGT | T,S 47 | NP_001295192.1 | |
NM_003799.2 | 381 | Missense Mutation | ACT,AGT | T,S 47 | NP_003790.1 | |
XM_005258162.1 | 381 | Missense Mutation | ACT,AGT | T,S 47 | XP_005258219.1 | |
XM_011525749.1 | 381 | Missense Mutation | ACT,AGT | T,S 47 | XP_011524051.1 | |
XM_011525750.2 | 381 | Missense Mutation | ACT,AGT | T,S 47 | XP_011524052.1 | |
XM_011525751.1 | 381 | Missense Mutation | ACT,AGT | T,S 47 | XP_011524053.1 | |
XM_011525753.1 | 381 | Intron | XP_011524055.1 | |||
XM_017026061.1 | 381 | Missense Mutation | ACT,AGT | T,S 47 | XP_016881550.1 |