Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATAGGGACCACCAAATACTGA[A/T]ATATGAAGACTCTATGACCAAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAPPC8 - trafficking protein particle complex 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014939.3 | 5119 | UTR 3 | NP_055754.2 | |||
XM_005258233.2 | 5119 | UTR 3 | XP_005258290.1 | |||
XM_005258234.2 | 5119 | UTR 3 | XP_005258291.1 | |||
XM_005258235.3 | 5119 | UTR 3 | XP_005258292.1 | |||
XM_006722420.3 | 5119 | Intron | XP_006722483.1 | |||
XM_011525884.2 | 5119 | UTR 3 | XP_011524186.1 | |||
XM_017025615.1 | 5119 | UTR 3 | XP_016881104.1 | |||
XM_017025616.1 | 5119 | UTR 3 | XP_016881105.1 | |||
XM_017025617.1 | 5119 | UTR 3 | XP_016881106.1 |