Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGATAACTGCACAACCTCTGGA[C/G]ACAAAGAGGCCGAATTGTCCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF397 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF397 - zinc finger protein 397 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135178.2 | 3899 | Intron | NP_001128650.1 | |||
NM_032347.2 | 3899 | Intron | NP_115723.1 | |||
XM_006722558.2 | 3899 | UTR 3 | XP_006722621.1 | |||
XM_011526229.2 | 3899 | UTR 3 | XP_011524531.1 | |||
XM_011526230.2 | 3899 | UTR 3 | XP_011524532.1 | |||
XM_011526231.2 | 3899 | UTR 3 | XP_011524533.1 | |||
XM_017026042.1 | 3899 | UTR 3 | XP_016881531.1 |
ZSCAN30 - zinc finger and SCAN domain containing 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112734.3 | 3899 | UTR 3 | NP_001106205.1 | |||
NM_001166012.2 | 3899 | UTR 3 | NP_001159484.1 | |||
NM_001288711.1 | 3899 | UTR 3 | NP_001275640.1 | |||
XM_005258183.4 | 3899 | UTR 3 | XP_005258240.1 | |||
XM_006722371.3 | 3899 | UTR 3 | XP_006722434.1 | |||
XM_011525789.2 | 3899 | UTR 3 | XP_011524091.1 | |||
XM_017025515.1 | 3899 | UTR 3 | XP_016881004.1 | |||
XM_017025516.1 | 3899 | UTR 3 | XP_016881005.1 | |||
XM_017025517.1 | 3899 | UTR 3 | XP_016881006.1 | |||
XM_017025518.1 | 3899 | UTR 3 | XP_016881007.1 | |||
XM_017025519.1 | 3899 | UTR 3 | XP_016881008.1 | |||
XM_017025520.1 | 3899 | UTR 3 | XP_016881009.1 | |||
XM_017025521.1 | 3899 | UTR 3 | XP_016881010.1 | |||
XM_017025522.1 | 3899 | UTR 3 | XP_016881011.1 |