Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTAAAGAGTTAGGAACATACCC[A/G]CTTTCATCTGCATCATAGAATAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMA8 - proteasome subunit alpha 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025096.1 | Intron | NP_001020267.1 | ||||
NM_001025097.1 | Intron | NP_001020268.1 | ||||
NM_001308188.1 | Intron | NP_001295117.1 | ||||
NM_144662.2 | Intron | NP_653263.2 | ||||
XM_011525818.1 | Intron | XP_011524120.1 |