Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCTTACACTTTTTGTTTAAATA[C/T]GAATTTAATTAAACAGAAAATTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALPK2 - alpha kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052947.3 | 6407 | UTR 3 | NP_443179.3 | |||
XM_011525801.2 | 6407 | Intron | XP_011524103.1 | |||
XM_011525802.2 | 6407 | UTR 3 | XP_011524104.1 | |||
XM_017025535.1 | 6407 | Intron | XP_016881024.1 | |||
XM_017025536.1 | 6407 | Intron | XP_016881025.1 | |||
XM_017025537.1 | 6407 | Intron | XP_016881026.1 | |||
XM_017025538.1 | 6407 | UTR 3 | XP_016881027.1 | |||
XM_017025539.1 | 6407 | UTR 3 | XP_016881028.1 |