Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAGTGGACTGTCTTGGCCAGAT[C/T]CCCTCCAGAAAACCAATATGGCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612197 | ||||||||||||||||||||
Literature Links: |
ABHD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABHD3 - abhydrolase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308256.1 | 1283 | Silent Mutation | GGA,GGG | G,G 328 | NP_001295185.1 | |
NM_001308257.1 | 1283 | Silent Mutation | GGA,GGG | G,G 186 | NP_001295186.1 | |
NM_138340.4 | 1283 | Silent Mutation | GGA,GGG | G,G 381 | NP_612213.2 | |
XM_011525847.2 | 1283 | Intron | XP_011524149.1 | |||
XM_017025571.1 | 1283 | Silent Mutation | GGA,GGG | G,G 186 | XP_016881060.1 | |
XM_017025572.1 | 1283 | Silent Mutation | GGA,GGG | G,G 186 | XP_016881061.1 | |
XM_017025573.1 | 1283 | Silent Mutation | GGA,GGG | G,G 186 | XP_016881062.1 | |
XM_017025574.1 | 1283 | Silent Mutation | GGA,GGG | G,G 186 | XP_016881063.1 |