Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGCGCTGTCCATTGCCATGGC[C/T]TCGCCCGTGGCCTACCACCAGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600377 | ||||||||||||||||||||
Literature Links: |
GALR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALR1 - galanin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001480.3 | 998 | Silent Mutation | GCC,GCT | A,A 167 | NP_001471.2 | |
XM_017025691.1 | 998 | Silent Mutation | GCC,GCT | A,A 167 | XP_016881180.1 |