Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAAATTCTGCAAAAGCAAGGAT[C/G]TAGAATTCCGAATTTTAGAAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMX3 - thioredoxin related transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019022.3 | 3885 | UTR 3 | NP_061895.3 | |||
XM_005266714.2 | 3885 | UTR 3 | XP_005266771.1 | |||
XM_011526033.2 | 3885 | UTR 3 | XP_011524335.1 | |||
XM_011526034.2 | 3885 | Intron | XP_011524336.1 | |||
XM_011526035.2 | 3885 | Intron | XP_011524337.1 |