Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGCCTTTCTCCCCATGGTCAG[C/T]CACACGTTCCACATGCGCACAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TXNDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TXNDC2 - thioredoxin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098529.1 | 659 | Silent Mutation | AGC,AGT | S,S 70 | NP_001091999.1 | |
NM_032243.5 | 659 | Silent Mutation | AGC,AGT | S,S 3 | NP_115619.4 | |
XM_017026041.1 | 659 | Silent Mutation | AGC,AGT | S,S 3 | XP_016881530.1 |