Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGATCCGGGACCTGAGTTCTT[G/T]GCGCACCAGGGAAACTCTGGCTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610347 | ||||||||||||||||||||
Literature Links: |
RPRD1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPRD1A - regulation of nuclear pre-mRNA domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303411.1 | 1019 | Missense Mutation | AAA,CAA | K,Q 241 | NP_001290340.1 | |
NM_001303412.1 | 1019 | Missense Mutation | AAA,CAA | K,Q 241 | NP_001290341.1 | |
NM_001303413.1 | 1019 | Intron | NP_001290342.1 | |||
NM_018170.4 | 1019 | Missense Mutation | AAA,CAA | K,Q 277 | NP_060640.2 | |
XM_011526048.1 | 1019 | Missense Mutation | AAA,CAA | K,Q 277 | XP_011524350.1 | |
XM_011526049.1 | 1019 | Missense Mutation | AAA,CAA | K,Q 241 | XP_011524351.1 | |
XM_011526050.1 | 1019 | Missense Mutation | AAA,CAA | K,Q 241 | XP_011524352.1 | |
XM_017025808.1 | 1019 | Intron | XP_016881297.1 |