Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCTATTAGAGCTAAAAGTATTA[C/T]TTTTAAAGTTATATGAGCAAGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPRD1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPRD1A - regulation of nuclear pre-mRNA domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303411.1 | 2031 | UTR 3 | NP_001290340.1 | |||
NM_001303412.1 | 2031 | Intron | NP_001290341.1 | |||
NM_001303413.1 | 2031 | Intron | NP_001290342.1 | |||
NM_018170.4 | 2031 | UTR 3 | NP_060640.2 | |||
XM_011526048.1 | 2031 | Intron | XP_011524350.1 | |||
XM_011526049.1 | 2031 | UTR 3 | XP_011524351.1 | |||
XM_011526050.1 | 2031 | UTR 3 | XP_011524352.1 | |||
XM_017025808.1 | 2031 | Intron | XP_016881297.1 |