Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTAAATTCAGATGGGCTTGGCC[A/G]TCAATAGTCTGTCCCAAGTTTATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSTPIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSTPIP2 - proline-serine-threonine phosphatase interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024430.3 | 1847 | UTR 3 | NP_077748.3 | |||
XM_011526253.2 | 1847 | UTR 3 | XP_011524555.1 | |||
XM_011526254.1 | 1847 | UTR 3 | XP_011524556.1 | |||
XM_011526256.1 | 1847 | UTR 3 | XP_011524558.1 |