Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTAGACAGTGGGGCGACAAAAA[A/G]CATTCCTGAATGGTGGATTCATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREB1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GREB1L - growth regulation by estrogen in breast cancer 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142966.1 | Intron | NP_001136438.1 | ||||
XM_006722547.2 | Intron | XP_006722610.1 | ||||
XM_011526179.2 | Intron | XP_011524481.1 | ||||
XM_017025988.1 | Intron | XP_016881477.1 | ||||
XM_017025989.1 | Intron | XP_016881478.1 | ||||
XM_017025990.1 | Intron | XP_016881479.1 | ||||
XM_017025991.1 | Intron | XP_016881480.1 | ||||
XM_017025992.1 | Intron | XP_016881481.1 | ||||
XM_017025993.1 | Intron | XP_016881482.1 | ||||
XM_017025994.1 | Intron | XP_016881483.1 | ||||
XM_017025995.1 | Intron | XP_016881484.1 | ||||
XM_017025996.1 | Intron | XP_016881485.1 |