Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAACCACCCTAGTAGTTCCGCT[C/G]CGAGGGCGGAAGTGCGCGTCTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616054 MIM: 608731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELP2 - elongator acetyltransferase complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242875.2 | 10 | UTR 5 | NP_001229804.1 | |||
NM_001242876.2 | 10 | UTR 5 | NP_001229805.1 | |||
NM_001242877.2 | 10 | UTR 5 | NP_001229806.1 | |||
NM_001242878.2 | 10 | UTR 5 | NP_001229807.1 | |||
NM_001242879.2 | 10 | UTR 5 | NP_001229808.1 | |||
NM_001324465.1 | 10 | UTR 5 | NP_001311394.1 | |||
NM_001324466.1 | 10 | UTR 5 | NP_001311395.1 | |||
NM_001324467.1 | 10 | UTR 5 | NP_001311396.1 | |||
NM_001324468.1 | 10 | UTR 5 | NP_001311397.1 | |||
NM_018255.3 | 10 | UTR 5 | NP_060725.1 |
SLC39A6 - solute carrier family 39 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099406.1 | 10 | Intron | NP_001092876.1 | |||
NM_012319.3 | 10 | Intron | NP_036451.3 | |||
XM_011525900.1 | 10 | Intron | XP_011524202.1 | |||
XM_011525901.1 | 10 | Intron | XP_011524203.1 |