Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCATGTCTCCGGGCAGCAGCGCA[C/T]GCTCCTTGGAGATGGGCAGGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616512 | ||||||||||||||||||||
Literature Links: |
RNF152 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF152 - ring finger protein 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173557.2 | 942 | Missense Mutation | CAT,CGT | H,R 110 | NP_775828.1 | |
XM_005266650.3 | 942 | Missense Mutation | CAT,CGT | H,R 110 | XP_005266707.1 | |
XM_005266652.3 | 942 | Missense Mutation | CAT,CGT | H,R 110 | XP_005266709.1 | |
XM_011525878.2 | 942 | Missense Mutation | CAT,CGT | H,R 110 | XP_011524180.1 | |
XM_011525879.2 | 942 | Missense Mutation | CAT,CGT | H,R 110 | XP_011524181.1 | |
XM_017025612.1 | 942 | Missense Mutation | CAT,CGT | H,R 110 | XP_016881101.1 | |
XM_017025613.1 | 942 | Missense Mutation | CAT,CGT | H,R 110 | XP_016881102.1 |