Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTGCAGCTTCCACTCCCTCCT[C/T]AGTGACCTCCACAAAGGCCTTGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600518 | ||||||||||||||||||||
Literature Links: |
SERPINB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINB4 - serpin family B member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002974.3 | 1159 | Missense Mutation | AAG,GAG | K,E 338 | NP_002965.1 | |
NM_175041.1 | 1159 | Missense Mutation | AAG,GAG | K,E 317 | NP_778206.1 | |
XM_011526138.1 | 1159 | Missense Mutation | AAG,GAG | K,E 338 | XP_011524440.1 |