Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAATGGAGGGATGTGGGCTGAG[A/C]TTTCAAACAGAATTAATGGTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615682 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINB11 - serpin family B member 11 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291278.1 | Intron | NP_001278207.1 | ||||
NM_001291279.1 | Intron | NP_001278208.1 | ||||
NM_080475.3 | Intron | NP_536723.2 | ||||
XM_011526251.1 | Intron | XP_011524553.1 |