Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGATCATAGAAGATATCCAGAGA[A/G]GGCTCCTGGACACAGTAGAGCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602932 | ||||||||||||||||||||
Literature Links: |
SMAD7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMAD7 - SMAD family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190821.1 | 431 | Silent Mutation | CCC,CCT | P,P 281 | NP_001177750.1 | |
NM_001190822.1 | 431 | Silent Mutation | CCC,CCT | P,P 67 | NP_001177751.1 | |
NM_001190823.1 | 431 | Silent Mutation | CCC,CCT | P,P 94 | NP_001177752.1 | |
NM_005904.3 | 431 | Silent Mutation | CCC,CCT | P,P 282 | NP_005895.1 |