Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGTTGGCGAAGCTTCCAGAACA[C/T]GCTGGAGGCTGCTCTCCGGGTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614544 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM69C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM69C - family with sequence similarity 69 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001044369.2 | 1250 | Missense Mutation | NP_001037834.2 | |||
XM_011525816.2 | 1250 | Missense Mutation | XP_011524118.1 | |||
XM_017025551.1 | 1250 | Missense Mutation | XP_016881040.1 |