Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCCCCAGCCTGTGTCTGCTGAA[A/G]TTAATGGGTGAAAAAGGTTGCACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604860 | ||||||||||||||||||||
Literature Links: |
LOC101927322 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927322 - uncharacterized LOC101927322 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MALT1 - MALT1 paracaspase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006785.3 | 540 | Silent Mutation | AAA,AAG | K,K 94 | NP_006776.1 | |
NM_173844.2 | 540 | Silent Mutation | AAA,AAG | K,K 94 | NP_776216.1 | |
XM_011525794.1 | 540 | Silent Mutation | AAA,AAG | K,K 94 | XP_011524096.1 |