Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATCTGTAGGTGACACATGAGT[A/G]ATCAAGCAGGTGCTCATTTACAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615115 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASXL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASXL3 - additional sex combs like 3, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030632.2 | Intron | NP_085135.1 | ||||
XM_005258356.1 | Intron | XP_005258413.1 | ||||
XM_011526205.2 | Intron | XP_011524507.1 | ||||
XM_011526206.2 | Intron | XP_011524508.1 | ||||
XM_011526209.1 | Intron | XP_011524511.1 | ||||
XM_011526210.1 | Intron | XP_011524512.1 | ||||
XM_011526212.1 | Intron | XP_011524514.1 | ||||
XM_011526213.1 | Intron | XP_011524515.1 | ||||
XM_017026012.1 | Intron | XP_016881501.1 | ||||
XM_017026013.1 | Intron | XP_016881502.1 | ||||
XM_017026014.1 | Intron | XP_016881503.1 |