Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTTGGATATCTTCGGGGACCTG[A/G]AGAAGATGAACAAGCGCCAGGTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SEC11C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC11C - SEC11 homolog C, signal peptidase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307941.1 | 159 | Missense Mutation | AAG,GAG | K,E 23 | NP_001294870.1 | |
NM_033280.3 | 159 | Missense Mutation | AAG,GAG | K,E 23 | NP_150596.1 | |
XM_011526260.2 | 159 | Intron | XP_011524562.1 | |||
XM_017026073.1 | 159 | Intron | XP_016881562.1 |