Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAAGCAGCATAAAAAGAATATT[C/T]TGCTCTCAATAAGCTTATGCTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINB10 - serpin family B member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005024.2 | Intron | NP_005015.1 | ||||
XM_011526027.1 | Intron | XP_011524329.1 | ||||
XM_011526028.1 | Intron | XP_011524330.1 | ||||
XM_017025793.1 | Intron | XP_016881282.1 | ||||
XM_017025794.1 | Intron | XP_016881283.1 |