Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCTATGGTTTTGTAGTAGTATG[C/T]TTTTTCCTCCTATATAAATAACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603049 MIM: 613473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXNL1 - thioredoxin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR7 - WD repeat domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015285.2 | Intron | NP_056100.2 | ||||
NM_052834.2 | Intron | NP_443066.2 | ||||
XM_006722431.2 | Intron | XP_006722494.1 | ||||
XM_011525888.1 | Intron | XP_011524190.1 | ||||
XM_017025682.1 | Intron | XP_016881171.1 | ||||
XM_017025683.1 | Intron | XP_016881172.1 |