Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGACGTCTTCACATTGGAAATC[A/G]TTCCACTGCCCAGCATAAATCAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607621 | ||||||||||||||||||||
Literature Links: |
COLEC12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COLEC12 - collectin subfamily member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130386.2 | 2227 | Silent Mutation | AAC,AAT | N,N 718 | NP_569057.1 | |
XM_011525741.2 | 2227 | Silent Mutation | AAC,AAT | N,N 701 | XP_011524043.1 |