Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTACTTCTTTTCGTGCTTCTTGG[C/T]TAACTCCCCAACGCCCAGTTGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAIN1 - A kinase (PRKA) anchor inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145194.1 | 2717 | Missense Mutation | ACC,GCC | T,A 63 | NP_001138666.1 | |
XM_011525728.2 | 2717 | Missense Mutation | ACC,GCC | T,A 70 | XP_011524030.1 |