Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATTTTCCCCCTAAACAGGCTG[C/G]CACTATGTCGAAGCTGCCCAAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607623 | ||||||||||||||||||||
Literature Links: |
C18orf8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C18orf8 - chromosome 18 open reading frame 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276342.1 | 474 | Missense Mutation | GCC,GGC | A,G 137 | NP_001263271.1 | |
NM_001318707.1 | 474 | Missense Mutation | GCC,GGC | A,G 28 | NP_001305636.1 | |
NM_001318708.1 | 474 | Missense Mutation | GCC,GGC | A,G 28 | NP_001305637.1 | |
NM_001318709.1 | 474 | Missense Mutation | GCC,GGC | A,G 137 | NP_001305638.1 | |
NM_013326.4 | 474 | Missense Mutation | GCC,GGC | A,G 185 | NP_037458.3 | |
XM_011525939.1 | 474 | Missense Mutation | GCC,GGC | A,G 28 | XP_011524241.1 | |
XM_017025717.1 | 474 | Missense Mutation | GCC,GGC | A,G 28 | XP_016881206.1 |
NPC1 - NPC intracellular cholesterol transporter 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000271.4 | 474 | Intron | NP_000262.2 | |||
XM_005258277.1 | 474 | Intron | XP_005258334.1 | |||
XM_005258278.4 | 474 | Intron | XP_005258335.1 | |||
XM_005258279.2 | 474 | Intron | XP_005258336.1 | |||
XM_006722479.3 | 474 | Intron | XP_006722542.1 | |||
XM_011526015.1 | 474 | Intron | XP_011524317.1 | |||
XM_017025784.1 | 474 | Intron | XP_016881273.1 | |||
XM_017025785.1 | 474 | Intron | XP_016881274.1 | |||
XM_017025786.1 | 474 | Intron | XP_016881275.1 | |||
XM_017025787.1 | 474 | Intron | XP_016881276.1 |