Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAACACAAGGTTCATCCAAATT[C/T]GTGTTTTTTTTTCACATATCACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NETO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NETO1 - neuropilin and tolloid like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201465.1 | 3143 | Intron | NP_001188394.1 | |||
NM_138966.3 | 3143 | UTR 3 | NP_620416.1 | |||
NM_138999.1 | 3143 | Intron | NP_620552.1 | |||
XM_005266774.4 | 3143 | Intron | XP_005266831.1 | |||
XM_017026017.1 | 3143 | UTR 3 | XP_016881506.1 | |||
XM_017026018.1 | 3143 | UTR 3 | XP_016881507.1 | |||
XM_017026019.1 | 3143 | UTR 3 | XP_016881508.1 | |||
XM_017026020.1 | 3143 | Intron | XP_016881509.1 | |||
XM_017026021.1 | 3143 | Intron | XP_016881510.1 | |||
XM_017026022.1 | 3143 | Intron | XP_016881511.1 | |||
XM_017026023.1 | 3143 | Intron | XP_016881512.1 | |||
XM_017026024.1 | 3143 | Intron | XP_016881513.1 |