Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAGTCTACTCCTCCCCCACAGGA[A/G]CCCACGATTTCAAATCCTCTTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613420 | ||||||||||||||||||||
Literature Links: |
KCTD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD1 - potassium channel tetramerization domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136205.2 | 797 | Silent Mutation | GGC,GGT | G,G 219 | NP_001129677.1 | |
NM_001142730.2 | 797 | Silent Mutation | GGC,GGT | G,G 827 | NP_001136202.1 | |
NM_001258221.1 | 797 | Silent Mutation | GGC,GGT | G,G 219 | NP_001245150.1 | |
NM_001258222.1 | 797 | Silent Mutation | GGC,GGT | G,G 227 | NP_001245151.1 | |
NM_198991.3 | 797 | Silent Mutation | GGC,GGT | G,G 219 | NP_945342.1 | |
XM_017025708.1 | 797 | Silent Mutation | GGC,GGT | G,G 219 | XP_016881197.1 |