Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGGTTGGCGCTCGGGTAGTGCG[C/T]GGCCCGGACTGGAAGTGGGGGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608677 | ||||||||||||||||||||
Literature Links: |
MIB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIB1 - mindbomb E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020774.3 | 851 | Silent Mutation | CGC,CGT | R,R 20 | NP_065825.1 | |
XM_011526098.1 | 851 | Intron | XP_011524400.1 | |||
XM_017025873.1 | 851 | Intron | XP_016881362.1 | |||
XM_017025874.1 | 851 | Silent Mutation | CGC,CGT | R,R 20 | XP_016881363.1 | |
XM_017025875.1 | 851 | Silent Mutation | CGC,CGT | R,R 20 | XP_016881364.1 |