Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCAGAGCATTGCCTTTTGGTTT[A/G]ACAAGCATTTTTAGAGGCCTACAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C18orf25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C18orf25 - chromosome 18 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008239.2 | Intron | NP_001008240.1 | ||||
NM_145055.3 | Intron | NP_659492.1 | ||||
XM_011525822.1 | Intron | XP_011524124.1 | ||||
XM_017025555.1 | Intron | XP_016881044.1 |