Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGAGTGTGTCCTCTAATGGATT[A/C]CTATACATAAAAAATGCACATGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 125645 | ||||||||||||||||||||
Literature Links: |
DSC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSC2 - desmocollin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004949.4 | 2970 | Nonsense Mutation | GAA,TAA | E,* 837 | NP_004940.1 | |
NM_024422.4 | 2970 | Intron | NP_077740.1 | |||
XM_005258206.4 | 2970 | Intron | XP_005258263.1 |