Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGATGCCAGATACTAATAGCA[C/T]AATCAATTTATCACTAAGCACTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606792 MIM: 615319 | ||||||||||||||||||||
Literature Links: |
HRH4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HRH4 - histamine receptor H4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143828.1 | 120 | Missense Mutation | ACA,ATA | T,I 7 | NP_001137300.1 | |
NM_001160166.1 | 120 | Missense Mutation | ACA,ATA | T,I 7 | NP_001153638.1 | |
NM_021624.3 | 120 | Missense Mutation | ACA,ATA | T,I 7 | NP_067637.2 | |
XM_011526133.1 | 120 | Missense Mutation | ACA,ATA | T,I 7 | XP_011524435.1 |
IMPACT - impact RWD domain protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |