Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCAGGTGGGCCTGTGTGGTGAA[A/T]CCTTTTGTGCACATTTCACAAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602126 | ||||||||||||||||||||
Literature Links: |
ZBTB14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB14 - zinc finger and BTB domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143823.2 | 1101 | Silent Mutation | GGA,GGT | G,G 313 | NP_001137295.1 | |
NM_001243702.1 | 1101 | Silent Mutation | GGA,GGT | G,G 313 | NP_001230631.1 | |
NM_001243704.1 | 1101 | Silent Mutation | GGA,GGT | G,G 313 | NP_001230633.1 | |
NM_003409.4 | 1101 | Silent Mutation | GGA,GGT | G,G 313 | NP_003400.2 |